1-Methylpyrrole 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21451-AE
B21451-AP
CAAAAB21451-0FEA
853.47
CAD
CAAAAB21451-0F
CAAAB21451-AE
CAAAB21451-AP
1-Methylpyrrole 99%
1-Methylpyrrole
Purity: 99%
CAS No.: 96-54-8
Molecular Formula: C5H7N
Formula Weight: 81.12
Flash Point: 15°(59°F)
Refractive Index: 1.4880
Melting Point: -57°
Boiling Point: 112-113°
Beilstein Reference: 104181
UN No.: 1993
MDL No.: MFCD00005345
Formula:
C₅H₇N Boiling Pt: 112…113 °C Melting Pt: –57 °C Density: 0.912 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005345 CAS Number: 96-54-8 EINECS: 202-513-7 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...