L(-)-Galactose 98%

Supplier: Thermo Scientific Chemicals

B21448-MC B21448-ME
CAAAB21448-MCEA 172.22 CAD
CAAAB21448-MC CAAAB21448-ME
L(-)-Galactose 98%
L(-)-Galactose

Purity: 98% CAS No.: 15572-79-9 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 163-165° Beilstein Reference: 1724622

Formula: C₆H₁₂O₆
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00063833
CAS Number: 15572-79-9
EINECS: 239-630-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR