Indane 95%

Supplier: Thermo Scientific Chemicals
Warning

B21442-0F B21442-AE B21442-AP
CAAAB21442-0FEA 1098.1 CAD
CAAAB21442-0F CAAAB21442-AE CAAAB21442-AP
Indane 95%
Indane

Purity: 95% CAS No.: 496-11-7 Molecular Formula: C9H10 Formula Weight: 118.18 Flash Point: 50°(122°F) Refractive Index: 1.5370 Melting Point: -51° Boiling Point: 175-177° Beilstein Reference: 1904376 Merck Reference: 14,4931 UN No.: 3295 MDL No.: MFCD00003795

Formula: C₉H₁₀
Boiling Pt: 175…177 °C
Melting Pt: –51 °C
Density: 0.962
Flash Pt: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003795
CAS Number: 496-11-7
EINECS: 207-814-7
UN: 1993
ADR: 3,III
Merck Index: 14,4931

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR