4-(Dimethylamino)antipyrine 98+%

Supplier: Thermo Scientific Chemicals
Warning

B21436-0B B21436-18 B21436-30
CAAAB21436-0BEA 1003.3 CAD
CAAAB21436-0B CAAAB21436-18 CAAAB21436-30
4-(Dimethylamino)antipyrine 98+%
4-(Dimethylamino)antipyrine

Purity: 98+% CAS No.: 58-15-1 Molecular Formula: C13H17N3O Formula Weight: 231.30 Odor: Odorless Melting Point: 105-109° UN No.: 2811 MDL No.: MFCD00003142

Formula: C₁₃H₁₇N₃O
Melting Pt: 105…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003142
CAS Number: 58-15-1
EINECS: 200-365-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,474

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR