Dipropyl ether 99%

Supplier: Thermo Scientific Chemicals
Danger

B21423-14 B21423-22
CAAAB21423-14EA 165.9 CAD
CAAAB21423-14 CAAAB21423-22
Dipropyl ether 99%
Dipropyl ether

Purity: 99% CAS No.: 111-43-3 Molecular Formula: (CH3CH2CH2)2O Formula Weight: 102.18 Flash Point: -28°(-18°F) Melting Point: -122° Boiling Point: 90-91° UN No.: 2384 MDL No.: MFCD00009376

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 90…91 °C
Melting Pt: –122 °C
Density: 0.747
Flash Pt: –28 °C (–18 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009376
CAS Number: 111-43-3
EINECS: 203-869-6
UN: 2384
ADR: 3 II
Merck Index: 12,08042

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR