2,3,4,5,6-Pentafluorotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

B21419-09 B21419-18
CAAAB21419-09EA 110.76 CAD
CAAAB21419-09 CAAAB21419-18
2,3,4,5,6-Pentafluorotoluene 99%
2,3,4,5,6-Pentafluorotoluene

Purity: 99% CAS No.: 771-56-2 Molecular Formula: C7H3F5 Formula Weight: 182.09 Flash Point: 34°(93°F) Refractive Index: 1.4030 Melting Point: -30° Boiling Point: 116-117° Beilstein Reference: 1911885 UN No.: 1993 MDL No.: MFCD00000298

Formula: C₇H₃F₅
MW: 182.09 g/mol
Boiling Pt: 116…117 °C
Melting Pt: –30 °C
Density: 1.439
Flash Pt: 34 °C (93 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000298
CAS Number: 771-56-2
EINECS: 212-233-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR