3-Bromophenetole 98%
Supplier: Thermo Scientific Chemicals
B21418-09
B21418-18
CAAAB21418-09EA
111.23
CAD
CAAAB21418-09
CAAAB21418-18
3-Bromophenetole 98%
3-Bromophenetole
Purity: 98%
CAS No.: 2655-84-7
Molecular Formula: C8H9BrO
Formula Weight: 201.06
Flash Point: 83°(182°F)
Refractive Index: 1.5480
Boiling Point: 231°
MDL No.: MFCD00274287
Formula:
C₈H₉BrO Boiling Pt: 231 °C Density: 1.41 Flash Pt: 83 °C(182 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00274287 CAS Number: 2655-84-7 EINECS: 220-186-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...