4-Nitropyridine-N-oxide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Nitropyridine N-oxide

B21404-14 B21404-22
CAAAAB21404-06EA 74.73 CAD
CAAAAB21404-06 CAAAB21404-14 CAAAB21404-22
4-Nitropyridine-N-oxide 97%
4-Nitropyridine N-oxide

Purity: 97% CAS No.: 1124-33-0 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: 158-163° Beilstein Reference: 124331 UN No.: 2811 MDL No.: MFCD00006206

Formula: C₅H₄N₂O₃
Boiling Pt: 397 °C (1013 hPa)
Melting Pt: 158…163 °C
Density: 1.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006206
CAS Number: 1124-33-0
EINECS: 214-395-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR