3,5-Difluorobenzoyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21400-06
B21400-14
CAAAB21400-06EA
107.12
CAD
CAAAB21400-06
CAAAB21400-14
3,5-Difluorobenzoyl chloride 98+%
3,5-Difluorobenzoyl chloride
Purity: 98+%
CAS No.: 129714-97-2
Molecular Formula: C7H3ClF2O
Formula Weight: 176.55
Flash Point: 69°(156°F)
Refractive Index: 1.5030
Boiling Point: 173-175°
Beilstein Reference: 4307685
UN No.: 3265
MDL No.: MFCD00010309
Formula:
C₇H₃ClF₂O Boiling Pt: 173…175 °C Density: 1.41 Flash Pt: 69 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010309 CAS Number: 129714-97-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...