Methyl-3-chlorobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21371-14
B21371-22
CAAAB21371-14EA
70.78
CAD
CAAAB21371-14
CAAAB21371-22
CAAAB21371-36
Methyl-3-chlorobenzoate 98%
Methyl 3-chlorobenzoate
Purity: 98%
CAS No.: 2905-65-9
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Flash Point: 104°(219°F)
Refractive Index: 1.5310
Melting Point: 20-22°
Boiling Point: 99-101°/12mm
Beilstein Reference: 2045126
Merck Reference: 14,2123
MDL No.: MFCD00000592
Formula:
C₈H₇ClO₂ Boiling Pt: 99…101 °C/12 mm Melting Pt: 20…22 °C Density: 1.227 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000592 CAS Number: 2905-65-9 EINECS: 220-814-1 Merck Index: 14,2123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...