Dodecylbenzene 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
n-Dodecylbenzene, 1-Phenyldodecane
B21362-06
B21362-14
B21362-22
CAAAB21362-06EA
56.56
CAD
CAAAB21362-06
CAAAB21362-14
CAAAB21362-22
Dodecylbenzene 97%
Dodecylbenzene
Purity: 97%
CAS No.: 123-01-3
Molecular Formula: C18H30
Formula Weight: 246.44
Flash Point: 124°(255°F)
Refractive Index: 1.4820
Melting Point: -7°
Boiling Point: 331°
Beilstein Reference: 1909107
MDL No.: MFCD00008974
Formula:
C₁₈H₃₀ Boiling Pt: 331 °C Melting Pt: –7 °C Density: 0.856 Flash Pt: 124 °C(255 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008974 CAS Number: 123-01-3 EINECS: 204-591-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...