1,3,5-Triphenylbenzene 99+%

Supplier: Thermo Scientific Chemicals

B21353-0B B21353-18 B21353-30
CAAAB21353-0BEA 1131.28 CAD
CAAAB21353-0B CAAAB21353-18 CAAAB21353-30
1,3,5-Triphenylbenzene 99+%
1,3,5-Triphenylbenzene

Purity: 99+% CAS No.: 612-71-5 Molecular Formula: C24H18 Formula Weight: 306.41 Melting Point: 172-174° Boiling Point: 460° MDL No.: MFCD00003060

Formula: C₂₄H₁₈
Boiling Pt: 460 °C
Melting Pt: 172…174 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003060
CAS Number: 612-71-5
EINECS: 210-318-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR