5-Methoxy-2-methylindole 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methoxy-2-methyl-1H-indole

B21348-03 B21348-06 B21348-14
CAAAB21348-03EA 62.25 CAD
CAAAB21348-03 CAAAB21348-06 CAAAB21348-14
5-Methoxy-2-methylindole 99+%
5-Methoxy-2-methylindole

Purity: 99+% CAS No.: 1076-74-0 Molecular Formula: C10H11NO Formula Weight: 161.20 Storage Temperature: 2°C to 8°C Melting Point: 85-88° MDL No.: MFCD00005620

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Boiling Pt: 145 °C (1.5 -rr)
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00005620
CAS Number: 1076-74-0
EINECS: 214-066-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR