3,5-Difluorobenzonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21347-06
B21347-14
CAAAB21347-06EA
64.62
CAD
CAAAB21347-06
CAAAB21347-14
3,5-Difluorobenzonitrile 99%
3,5-Difluorobenzonitrile
Purity: 99%
CAS No.: 64248-63-1
Molecular Formula: C7H3F2N
Formula Weight: 139.11
Melting Point: 83-88°
Boiling Point: 160°
UN No.: 3439
MDL No.: MFCD00010311
Formula:
C₇H₃F₂N Boiling Pt: 160 °C Melting Pt: 83…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010311 CAS Number: 64248-63-1 EINECS: 264-752-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...