2,5-Dichloro-p-xylene 98%
Supplier: Thermo Scientific Chemicals
B21344-14
B21344-22
CAAAB21344-14EA
112.02
CAD
CAAAB21344-14
CAAAB21344-22
2,5-Dichloro-p-xylene 98%
2,5-Dichloro-p-xylene
Purity: 98%
CAS No.: 1124-05-6
Molecular Formula: C8H8Cl2
Formula Weight: 175.06
Melting Point: 68-70°
Boiling Point: 221-223°
MDL No.: MFCD00000610
Formula:
C₈H₈Cl₂ MW: 175.06 Boiling Pt: 222 Melting Pt: 71 Density: 1.39 Storage Temperature: Ambient |
MDL Number:
MFCD00000610 CAS Number: 1124-05-6 EINECS: 214-387-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...