3-Bromo-4-hydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals

B21338-09 B21338-18
CAAAB21338-09EA 93.38 CAD
CAAAB21338-09 CAAAB21338-18
3-Bromo-4-hydroxybenzoic acid 98%
3-Bromo-4-hydroxybenzoic acid

Purity: 98% CAS No.: 14348-41-5 Molecular Formula: C7H5BrO3 Formula Weight: 217.02 Melting Point: 176-178° MDL No.: MFCD00017547

Formula: C₇H₅BrO₃
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00017547
CAS Number: 14348-41-5
EINECS: 238-299-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR