2-Amino-5-nitrothiazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21335-14
B21335-22
CAAAB21335-14EA
54.35
CAD
CAAAB21335-14
CAAAB21335-22
2-Amino-5-nitrothiazole 97%
2-Amino-5-nitrothiazole
Purity: 97%
CAS No.: 121-66-4
Molecular Formula: C3H3N3O2S
Formula Weight: 145.14
Melting Point: ca 202° dec.
Beilstein Reference: 126797
Merck Reference: 14,457
MDL No.: MFCD00005326
Formula:
C₃H₃N₃O₂S Melting Pt: ∼ 202 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005326 CAS Number: 121-66-4 EINECS: 204-490-9 Merck Index: 14,457 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...