α,4-Dibromo-2-fluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Bromo-1-(bromomethyl)-2-fluorobenzene, 4-Bromo-2-fluorobenzyl bromide
B21331-06
B21331-14
B21331-22
CAAAB21331-06EA
66.2
CAD
CAAAB21331-06
CAAAB21331-14
CAAAB21331-22
α,4-Dibromo-2-fluorotoluene 98%
α,4-Dibromo-2-fluorotoluene
Purity: 98%
CAS No.: 76283-09-5
Molecular Formula: C7H5Br2F
Formula Weight: 267.92
Melting Point: 33-37°
Boiling Point: 126°/9mm
UN No.: 3261
MDL No.: MFCD00055467
Formula:
C₇H₅Br₂F MW: 267.92 g/mol Boiling Pt: 126 °C/9 mm Melting Pt: 33…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055467 CAS Number: 76283-09-5 EINECS: 278-412-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...