2-Nitrobenzhydrazide 98%
Supplier: Thermo Scientific Chemicals
B21330-06
B21330-14
CAAAB21330-06EA
63.99
CAD
CAAAB21330-06
CAAAB21330-14
CAAAB21330-22
2-Nitrobenzhydrazide 98%
2-Nitrobenzhydrazide
Purity: 98%
CAS No.: 606-26-8
Molecular Formula: C7H7N3O3
Formula Weight: 181.15
Melting Point: 119-123°
Beilstein Reference: 394754
Formula:
C₇H₇N₃O₃ Melting Pt: 119…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007598 CAS Number: 606-26-8 EINECS: 210-110-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...