1,4-Diaminobutane 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Putrescine, 1,4-Butanediamine, Tetramethylendiamine

B21316-36 B21316-14 B21316-22
CAAAAB21316-36EA 413.96 CAD
CAAAAB21316-36 CAAAB21316-14 CAAAB21316-22
1,4-Diaminobutane 98+%
1,4-Diaminobutane

Purity: 98+% CAS No.: 110-60-1 Molecular Formula: C4H12N2 Formula Weight: 88.15 Flash Point: 63°(145°F) Refractive Index: 1.4569 Melting Point: 25-28° Boiling Point: 158-160° Beilstein Reference: 605282 Merck Reference: 14,7947 UN No.: 2928 MDL No.: MFCD00008235

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 158…160 °C
Melting Pt: 25…28 °C
Density: 0.877
Flash Pt: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008235
CAS Number: 110-60-1
EINECS: 203-782-3
UN: 2923
ADR: 8,III
Merck Index: 14,7947

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR