1,2-Dichlorobutane 97+%

Supplier: Thermo Scientific Chemicals
Warning

B21307-03 B21307-06
CAAAB21307-03EA 28.6 CAD
CAAAB21307-03 CAAAB21307-06
1,2-Dichlorobutane 97+%
1,2-Dichlorobutane

Purity: 97+% CAS No.: 616-21-7 Molecular Formula: C4H8Cl2 Formula Weight: 127.01 Flash Point: 30°(86°F) Refractive Index: 1.4450 Boiling Point: 124° Beilstein Reference: 1731376 UN No.: 1993 MDL No.: MFCD00013676

Formula: ClH₂CCHClCH₂CH₃
MW: 127.01 g/mol
Boiling Pt: 124 °C
Density: 1.112
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013676
CAS Number: 616-21-7
EINECS: 210-469-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR