Diisopropyl sulfide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isopropyl sulfide, Isopropyl sulphide

B21302-09 B21302-18
CAAAB21302-09EA 65.25 CAD
CAAAB21302-09 CAAAB21302-18
Diisopropyl sulfide 99%
Diisopropyl sulphide

Purity: 99% CAS No.: 625-80-9 Molecular Formula: C6H14S Formula Weight: 118.24 Flash Point: 7°(44°F) Refractive Index: 1.4385 Odor: Stench Boiling Point: 119-120° Beilstein Reference: 1731257 UN No.: 1993 MDL No.: MFCD00008895

Formula: C₆H₁₄S
MW: 118.24 g/mol
Boiling Pt: 67
Melting Pt: -69.1
Density: 0.814
Flash Pt: 7 °C (44 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008895
CAS Number: 625-80-9
EINECS: 210-911-7
UN: 1993
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR