2-Chloro-6-fluorobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

B21294-06 B21294-14
CAAAB21294-06EA 48.03 CAD
CAAAB21294-06 CAAAB21294-14 CAAAB21294-22
2-Chloro-6-fluorobenzamide 98%
2-Chloro-6-fluorobenzamide

Purity: 98% CAS No.: 66073-54-9 Molecular Formula: C7H5ClFNO Formula Weight: 173.58 Melting Point: 139-142° MDL No.: MFCD00055436

Formula: C₇H₅ClFNO
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00055436
CAS Number: 66073-54-9
EINECS: 266-123-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR