N-(Hydroxymethyl)phthalimide 97%
Supplier: Thermo Scientific Chemicals
B21292-14
B21292-22
B21292-36
CAAAB21292-14EA
45.5
CAD
CAAAB21292-14
CAAAB21292-22
CAAAB21292-36
N-(Hydroxymethyl)phthalimide 97%
N-(Hydroxymethyl)phthalimide
Purity: 97%
CAS No.: 118-29-6
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Melting Point: 140-147°
Beilstein Reference: 140946
Formula:
C₉H₇NO₃ Boiling Pt: 415 °C (1013 hPa) Melting Pt: 140…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005899 CAS Number: 118-29-6 EINECS: 204-241-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...