5-Iodosalicylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-5-iodobenzoic acid

B21291-36 B21291-14 B21291-22
CAAAAB21291-36EA 788.42 CAD
CAAAAB21291-36 CAAAB21291-14 CAAAB21291-22
5-Iodosalicylic acid 97%
5-Iodosalicylic acid

Purity: 97% CAS No.: 119-30-2 Molecular Formula: C7H5IO3 Formula Weight: 264.02 Melting Point: 194-199° Beilstein Reference: 2090443

Formula: C₇H₅IO₃
MW: 264.02 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 194…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00002458
CAS Number: 119-30-2
EINECS: 204-313-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR