2,6-Dichloropurine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21289-03
B21289-06
CAAAAB21289-MDEA
55.68
CAD
CAAAAB21289-MD
CAAAB21289-03
CAAAB21289-06
2,6-Dichloropurine 97%
2,6-Dichloropurine
Purity: 97%
CAS No.: 5451-40-1
Molecular Formula: C5H2Cl2N4
Formula Weight: 189.01
Melting Point: ca 184-186° dec.
Beilstein Reference: 9354
Formula:
C₅H₂Cl₂N₄ Melting Pt: ∼ 184…186 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00077725 CAS Number: 5451-40-1 EINECS: 226-681-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...