Methyl-2-bromo-5-nitrobenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21288-06
B21288-14
CAAAB21288-06EA
113.13
CAD
CAAAB21288-06
CAAAB21288-14
Methyl-2-bromo-5-nitrobenzoate 98+%
Methyl 2-bromo-5-nitrobenzoate
Purity: 98+%
CAS No.: 6942-36-5
Molecular Formula: C8H6BrNO4
Formula Weight: 260.05
Melting Point: 79-82°
Beilstein Reference: 2617575
Formula:
C₈H₆BrNO₄ Melting Pt: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010867 CAS Number: 6942-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...