2,5-Dibromo-1-fluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21281-14
B21281-22
CAAAB21281-14EA
122.77
CAD
CAAAB21281-14
CAAAB21281-22
2,5-Dibromo-1-fluorobenzene 98%
2,5-Dibromo-1-fluorobenzene
Purity: 98%
CAS No.: 1435-52-5
Molecular Formula: C6H3Br2F
Formula Weight: 253.91
Flash Point: 101°(213°F)
Melting Point: 33-35°
Boiling Point: 216°
Beilstein Reference: 3236451
MDL No.: MFCD00010603
Formula:
C₆H₃Br₂F Boiling Pt: 216 °C Melting Pt: 33…35 °C Flash Pt: 101 °C(213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010603 CAS Number: 1435-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...