Isoquinoline 97%

Supplier: Thermo Scientific Chemicals
Danger

B21279-22 B21279-36
CAAAAB21279-0EEA 1464.75 CAD
CAAAAB21279-0E CAAAB21279-22 CAAAB21279-36
Isoquinoline 97%
Isoquinoline

Purity: 97+% CAS No.: 119-65-3 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 107°(225°F) Refractive Index: 1.6245 Odor: Pungent pH: 7.5 Melting Point: 24-27° Boiling Point: 241-243° Beilstein Reference: 107549 Merck Reference: 14,5222 UN No.: 2811 MDL No.: MFCD00006898

Formula: C₉H₇N
Boiling Pt: 241…243 °C
Melting Pt: 24…27 °C
Density: 1.090
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006898
CAS Number: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,5222

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR