Dibenzyl diselenide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21278-14
B21278-03
B21278-06
CAAAAB21278-14EA
451.88
CAD
CAAAAB21278-14
CAAAB21278-03
CAAAB21278-06
Dibenzyl diselenide 95%
Dibenzyl diselenide
Purity: 95%
CAS No.: 1482-82-2
Molecular Formula: C6H5CH2Se-SeCH2C6H5
Formula Weight: 340.18
Melting Point: 91-93°
Beilstein Reference: 1877745
UN No.: 3283
MDL No.: MFCD00004767
Formula:
C₁₄H₁₄Se₂ Melting Pt: 91…93 °C Density: 1.67 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004767 CAS Number: 1482-82-2 UN: 3283 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...