3-Formylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isophthalaldehydic acid, 3-Carboxybenzaldehyde

B21277-14 B21277-03 B21277-06
CAAAAB21277-14EA 1276.64 CAD
CAAAAB21277-14 CAAAB21277-03 CAAAB21277-06
3-Formylbenzoic acid 97%
3-Formylbenzoic acid

Purity: 97% CAS No.: 619-21-6 Molecular Formula: C8H6O3 Formula Weight: 150.13 Melting Point: 173-175° Beilstein Reference: 2206413

Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00039575
CAS Number: 619-21-6
EINECS: 210-585-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR