4-Chloro-m-xylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21275-09
B21275-18
CAAAB21275-09EA
82.32
CAD
CAAAB21275-09
CAAAB21275-18
4-Chloro-m-xylene 97%
4-Chloro-m-xylene
Purity: 97%
CAS No.: 95-66-9
Molecular Formula: C8H9Cl
Formula Weight: 140.61
MDL No.: MFCD00060644
Formula:
C₈H₉Cl Storage Temperature: Ambient |
MDL Number:
MFCD00060644 CAS Number: 95-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...