2-Bromo-4,6-xylidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromo-2,4-dimethylaniline, 6-Bromo-2,4-xylidine, 2-Bromo-4,6-dimethylaniline

B21272-14 B21272-09 B21272-18
CAAAAB21272-14EA 167.48 CAD
CAAAAB21272-14 CAAAB21272-09 CAAAB21272-18
2-Bromo-4,6-xylidine 98%
2-Bromo-4,6-xylidine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 75…79 °C (5 mmHg)
Melting Pt: 42…46 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00047826
CAS Number: 41825-73-4
EINECS: 246-337-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR