3-Propylphenol 98%

Supplier: Thermo Scientific Chemicals
Danger

B21266-06 B21266-14
CAAAB21266-06EA 144.1 CAD
CAAAB21266-06 CAAAB21266-14 CAAAB21266-22
3-Propylphenol 98%
3-Propylphenol

Purity: 98% CAS No.: 621-27-2 Molecular Formula: C9H12O Formula Weight: 136.19 Refractive Index: 1.5250 Boiling Point: 229-231° UN No.: 3145 MDL No.: MFCD01632130

Formula: C₉H₁₂O
Boiling Pt: 229…231 °C
Storage Temperature: Ambient
MDL Number: MFCD01632130
CAS Number: 621-27-2
EINECS: 210-675-5
UN: 3145
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR