2'-Hydroxyacetanilide 97%

Supplier: Thermo Scientific Chemicals
Warning

B21264-14 B21264-22
CAAAB21264-14EA 50.09 CAD
CAAAB21264-14 CAAAB21264-22
2'-Hydroxyacetanilide 97%
2'-Hydroxyacetanilide

Purity: 97% CAS No.: 614-80-2 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 205-210° Beilstein Reference: 607592

Formula: C₈H₉NO₂
Melting Pt: 205…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00002181
CAS Number: 614-80-2
EINECS: 210-396-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR