4-tert-Butylbenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-(tert-Butyl)phenyl)methanol, (4-tert-Butylphenyl)methanol

B21260-06 B21260-14 B21260-22
CAAAB21260-06EA 70.31 CAD
CAAAB21260-06 CAAAB21260-14 CAAAB21260-22
4-tert-Butylbenzyl alcohol 98%
4-tert-Butylbenzyl alcohol

Purity: 98% CAS No.: 877-65-6 Molecular Formula: C11H16O Formula Weight: 164.25 Flash Point: 128°(262°F) Refractive Index: 1.5170 Boiling Point: 139-140°/20mm Beilstein Reference: 1860665 MDL No.: MFCD00004662

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 139…140 °C/20 mm
Density: 0.928
Flash Pt: 128 °C(262 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004662
CAS Number: 877-65-6
EINECS: 212-894-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR