2,6-Dichloro-3-nitropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

B21255-09 B21255-18 B21255-30
CAAAB21255-09EA 91.32 CAD
CAAAB21255-09 CAAAB21255-18 CAAAB21255-30
2,6-Dichloro-3-nitropyridine 97%
2,6-Dichloro-3-nitropyridine

Purity: 97% CAS No.: 16013-85-7 Molecular Formula: C5H2Cl2N2O2 Formula Weight: 192.99 Flash Point: 110°(230°F) Melting Point: 62-65° MDL No.: MFCD00006234

Formula: C₅H₂Cl₂N₂O₂
MW: 192.99
Melting Pt: 62…65 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006234
CAS Number: 16013-85-7
EINECS: 240-151-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR