3-Fluorophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21247-14
B21247-03
B21247-06
CAAAAB21247-14EA
701.52
CAD
CAAAAB21247-14
CAAAB21247-03
CAAAB21247-06
3-Fluorophenylboronic acid 97%
3-Fluorophenylboronic acid
Purity: 97%
CAS No.: 768-35-4
Molecular Formula: C6H6BFO2
Formula Weight: 139.92
Melting Point: 212-215°
Beilstein Reference: 3030632
Formula:
C₆H₆BFO₂ Melting Pt: 212…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236042 CAS Number: 768-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...