2-Octynoic acid methyl ester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 2-octynoate, Methyl oct-2-ynoate
B21242-14
B21242-22
CAAAB21242-14EA
58.46
CAD
CAAAB21242-14
CAAAB21242-22
2-Octynoic acid methyl ester 98%
2-Octynoic acid methyl ester
Purity: 98%
CAS No.: 111-12-6
Molecular Formula: CH3(CH2)4C≡CCO2CH3
Formula Weight: 154.20
Flash Point: 88°(192°F)
Refractive Index: 1.4460
Boiling Point: 216-218°
Beilstein Reference: 1756887
MDL No.: MFCD00009530
Formula:
C₉H₁₄O₂ MW: 154.21 g/mol Boiling Pt: 216…218 °C Density: 0.920 Flash Pt: 88 °C(192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009530 CAS Number: 111-12-6 EINECS: 203-836-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...