2,4-Diamino-6-hydroxypyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Aminoisocytosin, 2,4-Diamino-6-pyrimidinol, 2,4-Diaminopyrimidin-6-ol

B21240-14 B21240-22 B21240-36
CAAAB21240-14EA 52.61 CAD
CAAAB21240-14 CAAAB21240-22 CAAAB21240-36
2,4-Diamino-6-hydroxypyrimidine 96%
2,4-Diamino-6-hydroxypyrimidine

Purity: 96% CAS No.: 56-06-4 Molecular Formula: C4H6N4O Formula Weight: 126.12 Melting Point: 285-286° dec. Merck Reference: 14,2981 MDL No.: MFCD00006098

Formula: C₄H₆N₄O
MW: 126.12 g/mol
Boiling Pt: 373 °C (1013 hPa)
Melting Pt: 285…286 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006098
CAS Number: 56-06-4
EINECS: 200-254-4
Merck Index: 14,2981

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR