2-Phenylethyl acetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Phenylethyl Acetate, 2-Phenethyl acetate, Phenethyl acetate

B21238-0B B21238-30
CAAAB21238-0BEA 169.06 CAD
CAAAB21238-0B CAAAB21238-30
2-Phenylethyl acetate 98%
2-Phenylethyl acetate

Purity: 98% CAS No.: 103-45-7 Molecular Formula: CH3CO2(CH2)2C6H5 Formula Weight: 164.21 Flash Point: 107°(225°F) Refractive Index: 1.4990 Odor: Fruit-like Melting Point: -31° Boiling Point: 231-233° Beilstein Reference: 638179 MDL No.: MFCD00008720

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 231…233 °C
Melting Pt: –31 °C
Density: 1.029
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008720
CAS Number: 103-45-7
EINECS: 203-113-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR