4-Heptanone 98%

Supplier: Thermo Scientific Chemicals
Warning

B21222-AE B21222-AK B21222-AU
CAAAAB21222-AEEA 76.79 CAD
CAAAAB21222-AE CAAAB21222-AK CAAAB21222-AU
4-Heptanone 98%
4-Heptanone

Purity: 98% CAS No.: 123-19-3 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 48°(118°F) Refractive Index: 1.4070 Melting Point: -33° Boiling Point: 143-144° Beilstein Reference: 1699049 Merck Reference: 14,3345 UN No.: 2710 MDL No.: MFCD00009403

Formula: C₇H₁₄O
Boiling Pt: 143…144 °C
Melting Pt: –33 °C
Density: 0.818
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009403
CAS Number: 123-19-3
EINECS: 204-608-9
UN: 2710
ADR: 3,III
Merck Index: 14,3345

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR