1-Naphthyl boronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Naphthaleneboronic acid, Naphthalene-1-boronic acid, Naphthalen-1-ylboronic acid

B21219-14 B21219-03 B21219-06
CAAAAB21219-14EA 511.92 CAD
CAAAAB21219-14 CAAAB21219-03 CAAAB21219-06
1-Naphthyl boronic acid 97%
1-Naphthyl boronic acid

Purity: 97% CAS No.: 13922-41-3 Molecular Formula: C10H9BO2 Formula Weight: 171.99 Melting Point: 204-210° Beilstein Reference: 2937504

Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Melting Pt: 259
Storage Temperature: Ambient
MDL Number: MFCD00019722
CAS Number: 13922-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR