(±)-2-Pentanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Methyl propyl carbinol, (±)-2-Hydroxy pentane, DL-2-Pentanol, (±)-Propyl methyl carbinol

B21217-0F B21217-AP
CAAAB21217-0FEA 391.84 CAD
CAAAB21217-0F CAAAB21217-AP
(±)-2-Pentanol 99%
(±)-2-Pentanol

Purity: 99% CAS No.: 6032-29-7 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 33°(91°F) Physical Form: Liquid Density: 0.811 Refractive Index: 1.4060 Odor: Characteristic Melting Point: -50° Boiling Point: 114-115° Beilstein Reference: 1718819 Merck Reference: 14,7119 UN No.: 1105 MDL No.: MFCD00004579

Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 114…115 °C
Melting Pt: –50 °C
Density: 0.811
Flash Pt: 33 °C (91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004579
CAS Number: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: 3 III
Merck Index: 14,7119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc37771666c375a7768be6819fbc64e80617a202&vid=cc73accd09f3b4e43eedad1fe603cf52c119ff25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR