D-Methionine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH

B21213-06 B21213-14 B21213-22
CAAAB21213-06EA 53.56 CAD
CAAAB21213-06 CAAAB21213-14 CAAAB21213-22
D-Methionine 99%
D-Methionine

Purity: 99% CAS No.: 348-67-4 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Melting Point: ca 280° dec. Beilstein Reference: 1722293 Merck Reference: 14,5975 Optical Rotation: -23° (c=2 in 2N HCl) MDL No.: MFCD00002622

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002622
CAS Number: 348-67-4
EINECS: 206-483-6
Merck Index: 14,5975

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR