1-Chloro-2-fluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21209-14
B21209-22
CAAAB21209-14EA
150.57
CAD
CAAAB21209-14
CAAAB21209-22
1-Chloro-2-fluorobenzene 98+%
1-Chloro-2-fluorobenzene
Purity: 98+%
CAS No.: 348-51-6
Molecular Formula: C6H4ClF
Formula Weight: 130.55
Flash Point: 31°(87°F)
Refractive Index: 1.5010
Melting Point: -43°
Boiling Point: 137-138°
Beilstein Reference: 1855301
UN No.: 1993
MDL No.: MFCD00000533
Formula:
C₆H₄ClF Boiling Pt: 137…138 °C Melting Pt: –43 °C Density: 1.242 Flash Pt: 31 °C(87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000533 CAS Number: 348-51-6 EINECS: 206-476-8 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...