(3-Mercaptopropyl)triethoxysilane 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Triethoxysilyl)propanethiol, 3-Mercaptopropyltriethoxysilane
B21191-14
B21191-22
CAAAB21191-14EA
80.42
CAD
CAAAB21191-14
CAAAB21191-22
(3-Mercaptopropyl)triethoxysilane 94%
(3-Mercaptopropyl)triethoxysilane
Purity: 94%
CAS No.: 14814-09-6
Molecular Formula: C9H22O3SSi
Formula Weight: 238.42
Flash Point: 88°(190°F)
Physical Form: Liquid
Density: 0.98
Refractive Index: 1.4370
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless
Odor: Unpleasant
Boiling Point: 210°
Beilstein Reference: 2039575
MDL No.: MFCD00053924
Formula:
HSH₂CCH₂CH₂Si(OCH₂CH₃)₃ Boiling Pt: 210 °C Density: 0.98 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053924 CAS Number: 14814-09-6 EINECS: 238-883-1 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...