(S)-(-)-1-Phenylethanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol
B21188-03
B21188-06
CAAAB21188-03EA
63.52
CAD
CAAAB21188-03
CAAAB21188-06
(S)-(-)-1-Phenylethanol 98+%
(S)-(-)-1-Phenylethanol
For synthesis of optically active products
Purity: 98+%
CAS No.: 1445-91-6
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 85°(185°F)
Refractive Index: 1.5280
Odor: Mild
Melting Point: 9-11°
Boiling Point: 97-99°/20mm
Beilstein Reference: 2039797
Optical Rotation: -45° (c=5 in methanol)
UN No.: 2937
MDL No.: MFCD00064264
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 97…99 °C/20 mm Melting Pt: 9…11 °C Density: 1.012 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064264 CAS Number: 1445-91-6 UN: 2937 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...