D-(-)-Aspartic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH

B21184-06 B21184-14 B21184-22
CAAAB21184-06EA 45.98 CAD
CAAAB21184-06 CAAAB21184-14 CAAAB21184-22
D-(-)-Aspartic acid 99%
D(-)-Aspartic acid

Purity: 99% CAS No.: 1783-96-6 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Melting Point: 300° dec. Beilstein Reference: 1723529 Merck Reference: 14,840 Optical Rotation: -25° (c=1 in 1N HCl) MDL No.: MFCD00063081

Formula: C₄H₇NO₄
MW: 133.1 g/mol
Melting Pt: 251 (dec.)
Density: 1.66
Storage Temperature: Ambient
MDL Number: MFCD00063081
CAS Number: 1783-96-6
EINECS: 217-234-6
Merck Index: 14,840

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR