2-Nonadecanone tech. 80%
Supplier: Thermo Scientific Chemicals
B21178-06
B21178-14
CAAAB21178-06EA
98.75
CAD
CAAAB21178-06
CAAAB21178-14
2-Nonadecanone tech. 80%
2-Nonadecanone
Purity: 80%
CAS No.: 629-66-3
Molecular Formula: C19H38O
Formula Weight: 282.51
Flash Point:
Formula:
C₁₉H₃₈O MW: 282.51 g/mol Boiling Pt: 344 °C (1013 hPa) Melting Pt: 54…56 °C Density: 0.83 g/cm³ (20 °C) Flash Pt: >100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026671 CAS Number: 629-66-3 EINECS: 211-102-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...